Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium tin oxide coated glass slide, rectangular

Catalog No. 63691610PAK Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 pak
25 pak
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-691-610PAK 10 pak
63-691-625PAK 25 pak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-691-610PAK Supplier Sigma Aldrich Supplier No. 63691610PAK
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films. ITO coated glass has a sheet resistance of16+¼/cm2.ITO coated glass slides were used as substrates for barium vanadate films,α- andα-Fe2O3 nanostructures. The product was used in a study of photoelectrical properties of spin coated polyphenylene films. Poly(N-vinylcarbazole) was spin coated onto the substrate to form a light emitting layer towards the fabrication of light emitting diode (LED).

Specifications

CAS 50926-11-9
Quantity 10 pak
Linear Formula In2O3 -+ (SnO2)x
MDL Number MFCD00171662
Synonym ITO; ITO coated slide, rectangular
Refractive Index n20/D 1.517 (lit.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.