Learn More
Abcam 640/660 nm Extracellular Vesicle Membrane Staining Kit, 100 Tests
Description
- Assay type: Cell-based
- Detection method: Flow cytometry-fluorescent
- Applications: Flow Cyt
- Sample types: Suspension cells, Adherent cells
- Reactive species: Mammals
- Results type: Quantitative
- Labeling time: < 4 hrs
- Hands-on time: 30 secs
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| No. of Reactions | 100 Tests |
| For Use With (Equipment) | Flow Cytometer |
| Alternate Part Number | AB314688 |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Target Molecule | Extracellular Vesicles |
| Detection Method | Fluorescence |
| Cross Reactivity | Mammals |
| Sample Type | Suspension cells, Adherent cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.