Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam 640/660 Nm Extracellular Vesicle Membrane Staining Kit, 500 Tests
Description
- Assay type: Cell-based
- Applications: Flow Cyt
- Detection method: Flow cytometry-fluorescent
- Sample types: Suspension cells, Adherent cells
- Results type: Quantitative
- Reactive species: Mammals
- Key features: Labeling in < 4 hrs with 30 secs hands-on time
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Extracellular Vesicles |
| Shipping Condition | Ambient |
| Reactivity | Mammals |
| Format | Kit |
| No. of Reactions | 500 Tests |
| Content And Storage | Store at -20°C. |
| For Use With (Equipment) | Flow Cytometer |
| Detection Method | Fluorescence |
| Cross Reactivity | Mammals |
| Sample Type | Suspension cells, Adherent cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?