Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trans-3-Phenyl-1-Propen-1-Ylboronic Acid

Catalog No. 6426061G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-260-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-260-61G Supplier Sigma Aldrich Supplier No. 6426061G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in: Carbonylative arylation of allenolsOxidative cross-coupling with (trifluoromethyl)trimethylsilaneSuzuki-Miyaura coupling reactionsthree-component reductive coupling with alkynesEnantioselective conjugation to vinyl 2-pyridylsulfonesContains varying amounts of anhydride

Specifications

CAS 136725-55-8
Melting Point 179°C to 186°C
Molecular Formula C4H9ClFN
Linear Formula C4H9ClFN
MDL Number MFCD04038717
Quantity 1 g
Molecular Weight (g/mol) 125.57 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.