Learn More
ABclonal Technology 647 anti-Human CD270/HVEM
SDP
Supplier:  ABclonal Technology A24418500T
Encompass_Preferred
This gene encodes a member of the TNF (tumor necrosis factor) receptor superfamily The encoded protein functions in signal transduction pathways that activate inflammatory and inhibitory T-cell immune response It binds herpes simplex virus (HSV) viral envelope glycoprotein D (gD) mediating its entry into cells Alternative splicing results in multiple transcript variants
Catalog No. 50-000-09165