Learn More
Vector Laboratories Goat Anti-Rabbit, DyLight 594 DI-1649-1.5
SDP
Supplier:  Vector Laboratories DI16491.5
Encompass_Preferred
Goat Anti-Rabbit IgG Antibody (H+L), DyLight 594, 1.5 mg, is an affinity-purified, ultrapure, high-affinity antibody that is thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. Unless otherwise specified, antibodies recognize both heavy and light chains (H+L). Optimally labeled with DyLight 594 to provide the brightest label for fluorescence microscopy. Supplied in solution. Excitation: 592 nm; Emission: 617 nm; Color: Red.
Catalog No. NC1601047
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.