Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 65 cal Arc Flash Kit with Voltage Gloves (12)
SDP

Catalog No. 170011820 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1820 Large
17-001-1810 Small
17-001-1815 Medium
17-001-1825 2X-Large
17-001-1830 3X-Large
17-001-1835 4X-Large
17-001-1844 5X-Large
17-001-1845 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1820 Supplier National Safety Apparel™ Supplier No. KIT4SC65LG12
Add to Cart
Add to Cart

Inherently flame-resistant so they won’t burn, support combustion, or melt, making them a smart choice for those who encounter high levels of heat on the job

  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Made in the USA

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Material DuPont™ Nomex™, Kevlar™
Arc Flash Rating 65 cal/cm²
Seam Type Stitched
Suspender Adjustable
Color Brown
Size Large
Closure Type FR Hook and Loop
Ankle Style Open
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.