Learn More
National Safety Apparel™ 65 Cal ArcGuard™ Nomex/Kevlar Arc Flash Leggings
SDP
Catalog No. 170012042 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2042 X-Large
17-001-2041 Large
17-001-2043 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-001-2042 Supplier National Safety Apparel™ Supplier No. L05KDQTXL
Add to Cart
Add to Cart
Nomex/Kevlar Arc flash leggings
  • Inherently flame resistant for FR protection that will NOT wash out
  • Breathable FR fabric will NOT melt, drip, or ignite
  • FR fabric Wrap-around styling with FR hook and loop closure
  • Arc Rating: 65 cal/cm²
  • Fabric: Multilayer DuPont™ Nomex™ and Kevlar™
Specifications
Size X-Large
Gender Unisex
Color Brown
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Arc Flash Rating 65 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Sterility Non-sterile