Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 650 nm Stop Solution for TMB Substrate, 100 mL

Catalog No. 502582488
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2488 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2488 Supplier Abcam Supplier No. AB171531100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abcam 650 nm Stop Solution for TMB Substrate.

  • Species Reactivity: Species independent

Specifications

Product Type ELISA Stop Solution
Quantity 100 mL
For Use With (Application) ELISA, Immunohistochemistry (IHC)
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.