Learn More
Abcam 650 nm Stop Solution for TMB Substrate, 1000 mL
Description
- Species Reactivity: Species independent
Specifications
Specifications
| Product Type | ELISA Stop Solution |
| Quantity | 1000 mL |
| For Use With (Application) | ELISA, Immunohistochemistry (IHC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.