Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nickel(II) chloride hexahydrate

Catalog No. 6545075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-450-75G 5 g
65-450-7100G 100 g
65-450-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 65-450-75G Supplier Sigma Aldrich Supplier No. 6545075G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nickel(II) chloride hexahydrate is a nickel salt that can be used as a catalyst. It is cost effective and can be used in a variety of industrial processes. It can also be used in cross-coupling reactions.

Specifications

CAS 7791-20-0
Molecular Formula Cl2Ni -+ 6H2O
Linear Formula NiCl2 -+ 6H2O
Quantity 5 g
MDL Number MFCD00149809
Synonym Nickel chloride hexahydrate 2; Nickel dichloride hexahydrate
Molecular Weight (g/mol) 237.69
Percent Purity 99.9% trace metals basis
Biological Activity Lungs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.