Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tris(Triphenylphosphinegold)Oxonium Tetrafluoroborate

Catalog No. 6651421G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-514-21G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 66-514-21G Supplier Sigma Aldrich Supplier No. 6651421G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in a variety of gold catalyzed reactions including propargyl Claisen rearrangements.

Specifications

CAS 37095-27-5
Molecular Formula C25H22Au2Cl2P2
Linear Formula C25H22Au2Cl2P2
Quantity 250 mg
MDL Number MFCD08705280
Synonym Oxotris((triphenylphosphine)gold) tetrafluoroborate; Oxotris(triphenylphosphine)trigold tetrafluoroborate; Tris[(triphenylphosphino)aurio(I)]oxonium tetrafluoroborate
Molecular Weight (g/mol) 849.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.