Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Delta-Trisphat Tetrabutylammonium Salt

Catalog No. 669997500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-999-7500MG 500 mg
66-999-7100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 66-999-7500MG Supplier Sigma Aldrich Supplier No. 669997500MG
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Delta-Trisphat Tetrabutylammonium Salt

Specifications

CAS 301687-57-0
Molecular Formula C18Cl12O6P -+ C16H36N
Linear Formula C18Cl12O6P -+ C16H36N
MDL Number MFCD10566957
Quantity 500 mg
Synonym [Tetrabutylammonium] [Delta-tris(tetrachloro-1,2-benzenediolato)phosphate(V)]
Molecular Weight (g/mol) 1011.06 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.