Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dimethylimidazolium Dimethyl Phosphate

Catalog No. 67144450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-144-450G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-144-450G Supplier Sigma Aldrich Supplier No. 67144450G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Dimethylimidazolium dimethyl phosphate is an imidazolium-based phosphoric ionic liquid that can be prepared by reacting 1-methylimidazole with trimethyl phosphate. The pretreatment of sugarcane bagasse with [MMIM][DMP] yields glucose and xylose via enzymatic saccharification.1,3-Dimethylimidazolium dimethyl phosphate ([MMIM][DMP]) can be used as a catalyst and a solvent for the preparation of ethyl 2-cyano-3-phenylpropenoate from benzaldehyde and ethyl cyanoacetate via Knoevenagel condensation. It can also be used to produce bioethanol from sugarcane bagasse by enzymatic hydrolysis.

Specifications

CAS 68737-65-5
Melting Point 39°C to 44°C
Molecular Formula C8H18N2
Linear Formula C8H18N2
MDL Number MFCD00671527
Quantity 1 g
Synonym (R,R)-(-)-N,N'-Dimethyl-1,2-diaminocyclohexane; trans-1,2-Bis(methylamino)cyclohexane
Molecular Weight (g/mol) 142.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.