Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,7,10,13,16,19,22,25,32,35,38,41,44,47,50,53-Hexadecaoxa-28,29-Dithiahexapentacontanedioic Acid

Catalog No. 671525100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-152-5100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-152-5100MG Supplier Sigma Aldrich Supplier No. 671525100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of orthogonally dual-clickable Janus nanoparticles via cyclic templating strategyPreparation of cyclic RGD functionalized gold nanoparticles for tumor targetingAsymmetric-functionalized nanoparticle with orthogonal clickable functionalitiesFunctional immobilization of small GTPase Rab6A on DNA-gold nanoparticles by using a site-specifically attached poly(ethylene glycol) linker and thiol-place exchange reactionDirect writing of metal nanoparticle films inside sealed microfluidic channels in relation to biofunctionalization with ligands for protein capture and fluorescence assay

Specifications

CAS 505092-86-4
Melting Point 101°C to 110°C
Molecular Formula C13H13F6IrO2
Linear Formula Ir(C8H12)(C5HF6O2)
MDL Number MFCD07782019
Quantity 1 g
Molecular Weight (g/mol) 507.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.