Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 680C91, Tryptophan 2,3-Dioxygenase Inhibitor, Solid, 25 mg

Catalog No. 502704914
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4914 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4914 Supplier Abcam Supplier No. AB14615325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, and substrate-competitive tryptophan 2,3-dioxygenase inhibitor.

  • Potent, selective, substrate competitive tryptophan 2,3-dioxygenase inhibitor (Ki = 51 nM).
  • Inhibits tryptophan catabolism.
  • Increases brain tryptophan and serotonin levels in vivo.
  • Molecular weight: 238.26 Da.
  • Form: Solid.
  • Constituents: 100% 6-fluoro-3-[(E)-2-pyridin-3-ylethenyl]-1H-indole.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Tryptophan 2,3-dioxygenase
Buffer 100% 6-fluoro-3-[(E)-2-pyridin-3-ylethenyl]-1H-indole
Content And Storage Store at -20°C.
Alternate Part Number 680C91
Form Solid
Shipping Condition Ambient
Substrate Tryptophan
Formulation 100% 6-fluoro-3-[(E)-2-pyridin-3-ylethenyl]-1H-indole
Quantity 25 mg
For Use With (Application) Tryptophan catabolism inhibition
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.