Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 680C91, Tryptophan 2,3-Dioxygenase Inhibitor, Solid, 5 mg

Catalog No. 502704913
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4913 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4913 Supplier Abcam Supplier No. AB1461535MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and selective, substrate competitive tryptophan 2,3-dioxygenase inhibitor.

  • Potency: Potent, selective, substrate competitive tryptophan 2,3-dioxygenase inhibitor (Ki = 51 nM)
  • Biological activity: Inhibits tryptophan catabolism and increases brain tryptophan and serotonin levels in vivo
  • Molecular weight: 238.26 Da
  • Composition: 100% 6-fluoro-3-[(E)-2-pyridin-3-ylethenyl]-1H-indole
  • Form: Solid
  • CAS number: 163239-22-3
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Shipping Condition Ambient
Target Molecule Tryptophan 2,3-Dioxygenase
Content And Storage Store at -20°C.
Formulation 100% 6-fluoro-3-[(E)-2-pyridin-3-ylethenyl]-1H-indole
Quantity 5 mg
For Use With (Application) Tryptophan 2,3-dioxygenase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.