Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Alphatec™ 1800 Standard Bound Model 111 Coveralls with Hood

Catalog No. 17999981 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-981 Medium
17-999-980 Small
17-999-982 Large
17-999-983 X-Large
17-999-985 3X-Large
17-999-986 4X-Large
17-999-987 5X-Large
17-999-984 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17-999-981 Supplier Ansell Supplier No. WH18B9211103
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.

Coveralls with hood provide entry level, low hazard liquid and particulate protection for a wide range of general industrial applications. Previously known as MICROCHEM™ by AlphaTec 1800.

  • Provides low hazard liquid spray, splash and particle protection
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic waist, wrists, and ankles
  • 2-way front zipper with storm flap
  • Bound seams, attached hood
  • Available in Small through 5X-Large sizes
  • Certifications EN 1149-5, AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid, Anti-static
Certifications/Compliance AATCC 76, ANSI/ISEA 101
Certifications/Compliance (Europe) EN 1149-5, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
Latex Free No
Model WH18-B-92-111
No. of Pockets None
For Use With (Application) General Maintenance, Paint Spraying, Powder Handling, Food
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.