Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Formylphenylboronic Acid Pinacol Ester

Catalog No. 6838175G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-381-75G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 68-381-75G Supplier Sigma Aldrich Supplier No. 6838175G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Formylphenylboronic acid pinacol ester can be used as a starting material to prepare:Profluorescentprobe applicable for the detection of H2O2and Fe or Cu ions in living cells.Boronic acid pinacol ester derived nickel porphyrin, which is used to synthesize molecular spin switches and electron transfer dyads.Fluorescein hydrizido 2-imidophenylboronic ester, a fluorescent sensor prochelator applicable in the detection of copper ions.

Specifications

CAS 374067-51-3
Molecular Formula C48H46O4P2Ru
Refractive Index n20/D 1.5143
Linear Formula C48H46O4P2Ru
MDL Number MFCD09753023
Quantity 100 mg
Synonym Diacetato[(R)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]ruthenium(II)
Molecular Weight (g/mol) 849.89 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.