Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [6,6]-Phenyl C71 butyric acid methyl ester, mixture of isomers

Catalog No. 684465100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-446-5100MG 100 mg
68-446-5500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-446-5100MG Supplier Sigma Aldrich Supplier No. 684465100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[6,6]-Phenyl C71butyric acid methyl ester, mixture of isomers ([70]PCBM), is a high fullerene analog to [60]PCBM, which has an enhanced absorption of light in the visible region. It can be used in the development of electrochemical devices with a high current density.[70]PCBM can be blended with a variety of electron donors, which include P3HT and MDMO-PPV, for the fabrication of polymeric solar cells with a high power conversion efficiency (PCE). It can also be used in the organic field effect transistor (OFET) based applications.

Specifications

CAS 609771-63-3
Molecular Formula C82H14O2
Linear Formula C82H14O2
Quantity 100 mg
Synonym 3 H-Cyclopropa[8,25 ] [5,6 ]fullerene-C70-D5h(6)-3 butanoic acid; 3 Phenyl-3 H-cyclopropa[8,25 ] [5,6 ]fullerene-C70-D5h(6)-3 butanoic acid, methyl ester; [70 ]PCBM
Molecular Weight (g/mol) 1030.99
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.