Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Dicyclohexylphosphino-2',4',6'-Triisopropylbiphenyl Gold(I) Bis(Trifluoromethanesulfonyl)Imide

Catalog No. 6916741G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-167-41G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 69-167-41G Supplier Sigma Aldrich Supplier No. 6916741G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold Catalysts - 21st Century 'Gold Rush'Catalyst for isomerization of allenyl carbinol esters, hydroxy- and methoxycyclization, cycloisomerisation of diynes

Specifications

CAS 42237-85-4
Melting Point 217°C to 221°C
Molecular Formula C7H6BrNO2
Linear Formula C7H6BrNO2
MDL Number MFCD00227745
Quantity 5 g
Synonym XPhos AuNTf2
Molecular Weight (g/mol) 216.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.