Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-Lactic acid

Catalog No. 69785250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-785-250ML 250 mL
69-785-25L 2.5 L
69-785-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 69-785-250ML Supplier Sigma Aldrich Supplier No. 69785250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DL-Lactic acid can be used to synthesize serine racemase (SR) inhibitors.

Specifications

CAS 50-21-5
Boiling Point 122°C (15 mmHg)
Molecular Formula C3H6O3
Linear Formula CH3CH(OH)COOH
MDL Number MFCD00004520
Synonym 2-Hydroxypropionic acid
Molecular Weight (g/mol) 90.08
Percent Purity ∼90% (T)
Density 1.209 g/mL (at 25°C (literature))
Quantity 250 mL
Refractive Index n20/D 1.425 (literature)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.