Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(P-Sulfonatophenyl)Phenylphosphine Dihydrate Dipotassium Salt

Catalog No. 698539250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-853-9250MG 250 mg
69-853-92G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-853-9250MG Supplier Sigma Aldrich Supplier No. 698539250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(P-Sulfonatophenyl)Phenylphosphine Dihydrate Dipotassium Salt

Specifications

CAS 308103-66-4
Melting Point 98°C to 102°C
Molecular Formula C18H13K2O6PS2 -+ 2H2O
Linear Formula C18H13K2O6PS2 -+ 2H2O
MDL Number MFCD00269831
Quantity 250 mg
Synonym BSPP; 4,4'-(Phenylphosphinidene)bis-benzenesulfonic acid dipotassium salt
Molecular Weight (g/mol) 534.62 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.