Learn More
Bioassay Systems QuantiFluo™ ALP Reagent. Stabilized 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent for detecting alkaline phosphatase ALP, e.g. in ELISA.
SDP
Catalog No. 501956334 Shop All Bioassay Systems Products
Encompass_Preferred
Stabilized 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent for detecting alkaline phosphatase ALP, e.g. in ELISA. This single reagent produces a highly fluorescent product (360/450 nm) when dephosphorylated by ALP. The fluorescent product can also be visualized under a UV light. Key Features: Stable single reagent. Highly sensitive fluorescence detection. Can be visualized under a UV light. Method: FL360/450nm. Samples: Biological. Species: All. Procedure: Assay takes 30 min. Kit size: 600 tests.
QuantiFluo™ ALP Reagent is a stable 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent formulated for detecting alkaline phosphatase ALP, e.g. in ELISA. This single reagent produces a highly fluorescent product (λex/em = 360/450 nm) when dephosphorylated by ALP. The fluorescent product can also be visualized under a UV light.
Catalog No. 50-195-6334 Supplier Bioassay Systems Supplier No. QFALP6ML
Stabilized 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent for detecting alkaline phosphatase ALP, e.g. in ELISA. This single reagent produces a highly fluorescent product (360/450 nm) when dephosphorylated by ALP. The fluorescent product can also be visualized under a UV light. Key Features: Stable single reagent. Highly sensitive fluorescence detection. Can be visualized under a UV light. Method: FL360/450nm. Samples: Biological. Species: All. Procedure: Assay takes 30 min. Kit size: 600 tests.
QuantiFluo™ ALP Reagent is a stable 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent formulated for detecting alkaline phosphatase ALP, e.g. in ELISA. This single reagent produces a highly fluorescent product (λex/em = 360/450 nm) when dephosphorylated by ALP. The fluorescent product can also be visualized under a UV light.