Learn More
Across International 6mm ID Round Easy-Retrieve Dry Pressing Die Set
SDP
Supplier:  Across International SDS6.K
Encompass_Preferred
A set of 6mm diameter (ID) hardened steel trapezoidal split sleeve dry pressing die set for pellet presses, ready for you to make 6mm diameter cylinder-shape pellets. These die sets are professionally built for you to work with samples that tend to stick to the inner surface of the sleeve, trapezoidal split inner sleeves with pellet ejector make sample releasing a snap.
Catalog No. 50-225-1339