Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(6R)-5,6,7,8-Tetrahydro-L-Biopterin Dihydrochloride , MP Biomedical

Catalog No. MP215362250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215362250 50 mg
MP215362225 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215362250 Supplier MP Biomedicals, Inc Supplier No. 0215362250
Add to Cart
Add to Cart

(6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is a co-factor for nitric oxide synthase.

  • (6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is necessary for vivo for the conversion of phenylalanine to tyrosine and for the production of the hormone epinephrine and of the monoamine neurotransmitters, serotonin, dopamine, and norepinephrine
  • It is also involved in apoptosis and other cellular events mediated by nitric oxide production and catalyzes the cleavage of alkylglycerol ethers
  • (6R)-BH4 has been used as a buffer component for assaying tyrosine hydroxylase enzyme activity in mouse brain tissues
  • Natural cofactor for phenylalanine hydroxylase, tyrosine hydroxylase, tryptophan hydroxylase, and nitric oxide synthetase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H17Cl2N5O3
Molecular Weight (g/mol) 314.17
InChI Key RKSUYBCOVNCALL-NTVURLEBSA-N
Synonym sapropterin dihydrochloride, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, biopten, kuvan, sapropterin hydrochloride, 6r-5,6,7,8-tetrahydro-l-biopterin dihydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl
PubChem CID 636369
ChEBI CHEBI:32120

Specifications

Color White
Packaging Amber Glass Bottle
Quantity 50 mg
Percent Purity 99.6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.