Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride, Thermo Scientific™

Catalog No. AC328870250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328870250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328870250 Supplier Thermo Scientific Chemicals Supplier No. 328870250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H17Cl2N5O3
Molecular Weight (g/mol) 314.167
InChI Key RKSUYBCOVNCALL-NTVURLEBSA-N
Synonym sapropterin dihydrochloride, biopten, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, kuvan, sapropterin hydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl, 6r-tetrahydrobiopterin hydrochloride
PubChem CID 636369
ChEBI CHEBI:32120
IUPAC Name (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride
SMILES CC(C(C1CNC2=C(N1)C(=O)N=C(N2)N)O)O.Cl.Cl

Specifications

Quantity 25 mg
Merck Index 15, 8505
Formula Weight 314.3
Chemical Name or Material (6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.