Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride

Catalog No. AC328875000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328875000 500 mg
AC328871000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328875000 Supplier Thermo Scientific Chemicals Supplier No. 328875000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 69056-38-8 | C9H15N5O3·2HCl | 314.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H15N5O3·2HCl
Molecular Weight (g/mol) 314.17
InChI Key RKSUYBCOVNCALL-NTVURLEBSA-N
Synonym sapropterin dihydrochloride, biopten, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, kuvan, sapropterin hydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl, 6r-tetrahydrobiopterin hydrochloride
PubChem CID 636369
ChEBI CHEBI:32120
IUPAC Name (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride
SMILES CC(C(C1CNC2=C(N1)C(=O)N=C(N2)N)O)O.Cl.Cl

Specifications

Melting Point 250.0°C to 255.0°C
Quantity 500 mg
Merck Index 15,8505
Solubility Information Solubility in water: soluble.
Formula Weight 314.17
Chemical Name or Material (6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.