Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(2-Hydroxyethyl)theophylline 98.0+%, TCI America™
SDP

Catalog No. H040225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H040225G 25 g
H0402500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H040225G Supplier TCI America Supplier No. H040225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 519-37-9
Molecular Formula C9H12N4O3
Molecular Weight (g/mol) 224.22
MDL Number MFCD00055055
InChI Key NWPRCRWQMGIBOT-UHFFFAOYSA-N
Synonym Etofylline
PubChem CID 1892
IUPAC Name 7-(2-hydroxyethyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C2=C(N(CCO)C=N2)C(=O)N(C)C1=O

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7-(2-Hydroxyethyl)theophylline
EINECSNumber (5)-1132
RTECSNumber XH5850000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.