Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone 98.0+%, TCI America™
SDP

Catalog No. B33331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33331G 1 g
B33335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33331G Supplier TCI America Supplier No. B33331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129722-34-5
Molecular Formula C13H16BrNO2
Molecular Weight (g/mol) 298.18
MDL Number MFCD06658540
InChI Key URHLNHVYMNBPEO-UHFFFAOYSA-N
Synonym 7-(4-Bromobutoxy)-1,2,3,4-tetrahydro-2-oxoquinoline
PubChem CID 10542064
IUPAC Name 7-(4-bromobutoxy)-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)OCCCCBr

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.