Learn More
Abcam 7-AAD Staining Solution, Cell-based, Flow Cyt, 400 Test
Description
- Assay type: Cell-based
- Applications: Flow Cyt
- Sample types: Suspension cells
- Molecular Weight: 1270.4 Da
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | 7-AAD |
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Flow Cytometer |
| Format | Solution |
| Form | Solution |
| Label or Dye | 7-AAD |
| No. of Reactions | 400 Tests |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.