Learn More
Medchemexpress LLC Acetyltaxol, 7-(P) | 92950-39-5 | MFCD09264627 | C49H53NO15 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13805610MG
Encompass_Preferred
7-Acetyl paclitaxel is a paclitaxel derivative that inhibits microtubule depolymerization and is used as a research tool to study microtubule dynamics and cytoskeleton-targeting compounds.
  • Microtubule depolymerization inhibitor useful for studying microtubule dynamics.
  • Demonstrated activity: reduces growth of J774.2 macrophages (IC50 ~60 nM).
  • Available in milligram quantities suitable for in vitro and analytical use.
  • Certificate of analysis provides lot-specific purity and storage instructions.
  • For research use only; not for human or veterinary applications.
Catalog No. 50-002-59400