Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminodesacetoxycephalosporanic Acid 98.0+%, TCI America™
SDP

Catalog No. A207525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A207525G 25 g
A20755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A207525G Supplier TCI America Supplier No. A207525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22252-43-3
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
MDL Number MFCD00151456
InChI Key NVIAYEIXYQCDAN-CLZZGJSISA-N
Synonym 7-aminodesacetoxycephalosporanic acid, 6r,7r-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7-aminodeacetoxycephalosporanic acid, 7-adca, unii-anm3msm8tn, anm3msm8tn, 7beta-amino-3-methyl-3-cephem-4-carboxylic acid, 7beta-amino-3-methylceph-3-em-4-carboxylic acid, 7-amino desacetoxy cephalosporanic acid, 7beta-aminodeacetoxycephalosporanic acid
PubChem CID 33498
ChEBI CHEBI:64984
IUPAC Name (6R,7R)-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)N)SC1)C(=O)O

Specifications

Melting Point 242°C
Color Yellow
Quantity 25 g
Formula Weight 214.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminodesacetoxycephalosporanic Acid
EINECSNumber (8)-0483
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.