Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminoheptanoic Acid 98.0+%, TCI America™
SDP

Catalog No. A031125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A031125G 25 g
A03111G 1 g
A03115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A031125G Supplier TCI America Supplier No. A031125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 929-17-9
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00008242
InChI Key XDOLZJYETYVRKV-UHFFFAOYSA-N
Synonym 7-aminoenanthic acid, heptanoic acid, 7-amino, omega-aminoenantic acid, aminoenanthic acid, amino-enanthylic acid, 7-amino-heptanoic acid, 7-aminohepentanoic acid, omega-aminoheptanoic acid, unii-b10hjx2t48, kyselina 7-aminoenanthova czech
PubChem CID 13580
IUPAC Name 7-aminoheptanoic acid
SMILES C(CCCN)CCC(=O)O

Specifications

Melting Point 189°C
Color White
Quantity 25 g
Formula Weight 145.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminoheptanoic Acid
RTECSNumber MJ1770000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.