Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Bromo[1,2,4]triazolo[1,5-a]pyridine

Catalog No. SYX004031G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX004031G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX004031G Supplier Sigma Aldrich Supplier No. SYX004031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reaction between chloropentafluorobenzene and strong n-bases in polar aprotic solvents and in the presence of water has been investigated. chloropentafluorobenzene on reaction with ammonia yields 4-chloro-2,3,5,6-tetrafluoroaniline and 2-chloro-3,4,5,6-tetrafluoroaniline.chloropentafluorobenzene has been used in the preparation of:
  • 1,2-difluoro-1,2-bis-(pentafluorophenyl)dichlorane via fluorination with elemental fluorine at 128c
  • 5-chloro-1-(difluorochloro)-2,3,4,5,6,6-hexafluoro-1,3-cyclohexadiene via reaction with chlorine trifluoride at 78c

Specifications

CAS 1053655-66-5
Molecular Formula C6H4BrN3
Linear Formula C6H4BrN3
Quantity 25 g
MDL Number MFCD10699200
Molecular Weight (g/mol) 198.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.