Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromo-5-chloro-8-hydroxyquinoline 98.0+%, TCI America™
SDP

Catalog No. B11901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11901G 1 g
B11905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11901G Supplier TCI America Supplier No. B11901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7640-33-7
Molecular Formula C9H5BrClNO
Molecular Weight (g/mol) 258.499
MDL Number MFCD00044601
InChI Key ADJQQSUBKRASQN-UHFFFAOYSA-N
PubChem CID 82095
IUPAC Name 7-bromo-5-chloroquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Cl)Br)O)N=C1

Specifications

Melting Point 181°C
Quantity 1 g
Formula Weight 258.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-5-chloro-8-hydroxyquinoline
RTECSNumber VC4585000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.