Learn More
Cayman Chemical 7-BromoheptnoIc AcId 5g
Supplier: Cayman Chemical 304205 G
An L-type calcium channel blocker inhibits calcium-induced contraction of rabbit ear artery (pA2 9 4) induces relaxation of calcium-induced contractions in isolated rat colon and bladder and guinea pig trachea (IC50s 6 7 6 and 7 8 nM respectively) induces negative inotropy in isolated guinea pig ventricular strips (IC50 110 nM) reduces mean blood pressure in spontaneously hypertensive rats (ED25 0 35 mg/kg) and in renal hypertensive dogs (ED25 0 22 mg/kg) inhibits copper-induced oxidation of isolated human LDL at 1 and 5 UM reduces the extension of aortic atheromatous lesions and decreases renal injury in ApoE-/- mice in a model of Western diet-induced atherosclerosis
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.