Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromooxindole 98.0+%, TCI America™
SDP

Catalog No. B50971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50971G 1 g
B5097200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50971G Supplier TCI America Supplier No. B50971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 320734-35-8
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD02179613
InChI Key WSUWXWBRIBGIQT-UHFFFAOYSA-N
Synonym 7-Bromo-2-indolinone
PubChem CID 2773291
IUPAC Name 7-bromo-1,3-dihydroindol-2-one
SMILES C1C2=C(C(=CC=C2)Br)NC1=O

Specifications

Melting Point 209°C
Quantity 1 g
Formula Weight 212.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromooxindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.