Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromoquinoline, 95%

Catalog No. AAH63987MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63987MD 250 mg
AAH6398703 1 g
AAH6398706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63987MD Supplier Thermo Scientific Chemicals Supplier No. H63987MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 4965-36-0 | C9H6BrN | 208.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4965-36-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD03695823
InChI Key XYBSZCUHOLWQQU-UHFFFAOYSA-N
Synonym quinoline, 7-bromo, 7-bromo-quinoline, pubchem12788, ksc238o9n, abbypharma ap-12-10883
PubChem CID 521259
IUPAC Name 7-bromoquinoline
SMILES C1=CC2=C(C=C(C=C2)Br)N=C1

Specifications

Quantity 250 mg
Formula Weight 208.06
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Bromoquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.