Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Deazapurine 98.0+%, TCI America™
SDP

Catalog No. D48201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48201G 1 g
D4820200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48201G Supplier TCI America Supplier No. D48201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 271-70-5
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD08234403
InChI Key JJTNLWSCFYERCK-UHFFFAOYSA-N
Synonym 7H-Pyrrolo[2,3-d]pyrimidine
PubChem CID 261591
IUPAC Name 7H-pyrrolo[2,3-d]pyrimidine
SMILES N1C=CC2=CN=CN=C12

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 1 g
Formula Weight 119.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Deazapurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.