Learn More
AdipoGen 7-Diethylamino-3-MD-ethyl-CC
SDP
Supplier:  AdipoGen CDXD0198M001
Encompass_Preferred
Chemical. CAS 156571-46-9. Formula C20H21N3O5. MW 383.4. Synthetic. Thiol-reactive fluorescent probe for protein labeling. MDCC has been used for preparing specific probes for inorganic phosphate.
Catalog No. 50-256-1198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.