Learn More
Abcam 7-Diethylamino-3-[N-(4-maleimidoethyl)carbamoyl]coumarin, Thiol-Reactive Fluorescent Probe, >97% Purity, Powder, 20 mg
Description
- Application: Thiol-reactive fluorescent probe for protein labeling
- Purity: >97%
- Molecular Weight: 383.4 g/mol
- Form: Powder
- CAS Number: 156571-46-9
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | Fluorescent Probe |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Proteins |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Product Type | Protein Labeling Reagent |
| Buffer | 100% 7-Diethylamino-3-[N-(2-maleimidoethyl)carbamoyl]coumarin |
| Form | Powder |
| Detection Method | Fluorescence |
| Quantity | 20 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.