Learn More
Abcam 7-Diethylamino-3-[N-(4-maleimidoethyl)carbamoyl]coumarin, Thiol-Reactive Fluorescent Probe, Powder, >97% Purity, 1 mg
Description
- Molecular Weight: 383.4 g/mol
- Purity: >97%
- Form: Powder
- Application: Thiol-reactive fluorescent probe for protein labeling
- Storage: Store at -20°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Thiols |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Label or Dye | Fluorescent Probe |
| Form | Powder |
| Product Type | Protein Labeling Reagent |
| Buffer | 100% 7-Diethylamino-3-[N-(2-maleimidoethyl)carbamoyl]coumarin |
| Content And Storage | Store at -20°C. |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.