Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(Diethylamino)coumarin 98.0+%, TCI America™
SDP

Catalog No. D33541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33541G 1 g
D3354200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33541G Supplier TCI America Supplier No. D33541G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20571-42-0
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.27
MDL Number MFCD00232946
InChI Key QXAMGWKESXGGNV-UHFFFAOYSA-N
PubChem CID 88598
IUPAC Name 7-(diethylamino)-2H-chromen-2-one
SMILES CCN(CC)C1=CC=C2C=CC(=O)OC2=C1

Specifications

Melting Point 89°C
Color Yellow
Quantity 1 g
Formula Weight 217.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Diethylamino)coumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.