Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 7-Diethylaminocoumarin-3-Carboxylic Acid N-Succinimidyl Ester, >95% Purity, Powder, 100 mg

Catalog No. 502583263
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3263 Supplier Abcam Supplier No. AB145264100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent labeling reagent for amine-containing biomolecules, featuring >95% purity.

  • Application: Fluorescent labeling reagent for amine-containing biomolecules, including amino acids prior to resolution by capillary zone electrophoresis.
  • Purity: >95%
  • Molecular Weight: 358.35 g/mol
  • Form: Powder
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Amine-containing Biomolecules
Substrate Amine-containing biomolecules
Shipping Condition Ambient
Label or Dye 7-Diethylaminocoumarin
For Use With (Equipment) Capillary Electrophoresis Systems
Product Type Protein Labeling Reagent
Content And Storage Store at -20°C.
Detection Location In-Solution Detection
Detection Method Fluorescence
Form Powder
Formulation 100% (2,5-dioxopyrrolidin-1-yl) 7-(diethylamino)-2-oxochromene-3-carboxylate
Quantity 100 mg
For Use With (Application) Fluorescent labeling of amine-containing biomolecules, labeling of amino acids prior to capillary zone electrophoresis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.