Learn More
Cayman Chemical 7-epI-10-Deacetyltaxol 5mg
SDP
Supplier:  Cayman Chemical 342965 MG
Encompass_Preferred
A coumarin with diverse biological activities reduces A23187-induced release of PGE2 and LTC4 in isolated mouse peritoneal macrophages active against S. aureus S. epidermidis B. subtilis S. lutea and V. cholera (MICs 0.5 0.75 0.75 0.75 and 0.5 mg/ml respectively) reduces formalin-induced paw licking and biting responses in mice at 3 and 7.5 mg/kg
Catalog No. 50-294-6177
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.