Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Ethoxybenzo[b]furan-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AAB2114303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2114303 1 g
AAB2114306 5 g
AAB2114314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2114303 Supplier Thermo Scientific Chemicals Supplier No. B2114303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206559-61-7
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00060710
InChI Key DQJVZIUEXPTEKX-UHFFFAOYSA-N
PubChem CID 736202
IUPAC Name 7-ethoxy-1-benzofuran-2-carboxylic acid
SMILES CCOC1=CC=CC2=C1OC(=C2)C(=O)O

Specifications

Melting Point 196°C to 199°C
Quantity 1 g
Synonym 7-ethoxybenzofuran-2-carboxylic acid, 7-ethoxybenzo b furan-2-carboxylic acid, acmc-1cirp, 2-benzofurancarboxylicacid,7-ethoxy, 2-benzofurancarboxylicacid, 7-ethoxy, 2-benzofurancarboxylic acid, 7-ethoxy, 2-benzofurancarboxylic acid, 7-ethoxy-9ci
Formula Weight 206.2
Percent Purity 97%
Chemical Name or Material 7-Ethoxybenzo[b]furan-2-carboxylic acid
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.