Learn More
AdipoGen 7-MeO-coumarin-3-carbonyl azid
SDP
Supplier:  AdipoGen CDXM0018M100
Encompass_Preferred
Chemical. CAS 97632-67-2. Formula C11H7N3O4. MW 220.18. Synthetic. Fluorescent reagent.
Catalog No. 50-256-0567
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.