Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-4-quinolinol 97.0+%, TCI America™
SDP

Catalog No. M19661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19661G 1 g
M19665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19661G Supplier TCI America Supplier No. M19661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82121-05-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00169015
InChI Key NQUPXNZWBGZRQX-UHFFFAOYSA-N
Synonym 4-Hydroxy-7-methoxyquinoline
PubChem CID 2831995
IUPAC Name 7-methoxy-1H-quinolin-4-one
SMILES COC1=CC2=C(C=C1)C(=O)C=CN2

Specifications

Melting Point 285°C
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-4-quinolinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.