Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-methoxy-benzofuran-2-carbaldehyde

Catalog No. CDS012855 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS012855 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS012855 Supplier Sigma Aldrich Supplier No. CDS01285525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-benzylethylenediamine participates in the one-pot synthesis of n,n,nα-trisubstituted guanidines
  • It undergoes condensation with dibenzoylmethane (1,3-diphenyl-1,3-propanedione) in stoichiometric ratio 1:1 to afford the corresponding schiff monobase.n-benzylethylenediamine may be used for the synthesis of n-benzyl-n,nα,nα-tris(tert-butyloxycarbonylmethyl)ethylenediamine and 3-benzyl-2-(phenyl-2-sulfonate)-2-imidazoline tetraheptylammonium salt

Specifications

CAS 88234-77-9
Molecular Formula C10H8O3
Linear Formula C10H8O3
MDL Number MFCD03984484
Quantity 25 mL
Molecular Weight (g/mol) 176.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.